Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ Chemical- and Flame-Resistant Jackets

Catalog No. 17987443E Shop All Ansell Products
Click to view available options
Color:
Blue
Red
Material:
CPC Nomex
CPC Polyester
Size:
Large
X-Large
2X-Large
This item is not returnable. View return policy
Gore™ ChemSplash™ fabric is vapor permeable and should not be used for protection against hazardous vapors, carcinogens, or other life-threatening materials. Nomex™ and Gore ChemSplash fabric are not always in stock. Call your Fisher Safety Customer Service Representative for current information.

Catalog No. 17-987-443E

Supplier: Ansell™ 66670XL

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Gore™ ChemSplash™ fabric is vapor permeable and should not be used for protection against hazardous vapors, carcinogens, or other life-threatening materials. Nomex™ and Gore ChemSplash fabric are not always in stock. Call your Fisher Safety Customer Service Representative for current information.

Lightweight and breathable CPC garments made with Gore™ ChemSplash™ fabric reduce overheating and fatigue while preventing liquid chemical splash penetration

Gore membrane is a bicomponent PTFE membrane that prevents liquid penetration but allows a vapor permeation, allowing perspiration to evaporate and cool the wearer.

  • Garments stitched with Gore thread
  • All seams are heat and pressure sealed with Gore-Seam™ tape to prevent liquid penetration
  • Fabrics are durable, flexible, tear and snag resistant, washable, and reusable

Jackets
  • Heavy-duty zipper
  • Double storm flap; stand-up collar; raglan sleeves
  • Hook-and-loop closures on storm flap, collar, and sleeves
  • Sized to be worn over clothing

Order Info

For chemical splash protection, choose CPC garments made with red polyester Gore ChemSplash fabric (7 oz. per sq. yd.). When flame protection is also required, choose garments made with blue aramid (Nomex™) Gore ChemSplash fabric (6 oz. per sq. yd.); when exposed to flame, it will self-extinguish when removed from the flame source, and not contribute to burn injury.

TRUSTED_SUSTAINABILITY

Specifications

Size X-Large
Material CPC Nomex
Color Blue
Closure Type Heavy-duty Zipper
Construction Trilaminate
Collar Type Stand-up
Size (English) Chest 58 in.
Size (Metric) Chest 147.3cm
For Use With (Application) Industries, bulk chemical handling and transport, chemical plants and transportation, chemical processing, Oil refining and petrochemicals, handling specialty chemicals
Length (English) 30 in.
Length (Metric) 76.2cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.