Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell EDGE™ 40-400 Porous Nitrile Coated Medium Duty Cotton Gloves

Catalog No. 19019828 Shop All Ansell Products
missing translation for 'orderingAttributeHoverText'
Size:
10
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'

Porous nitrile coating helps channel oil away from the glove's surface for a firmer grip in slightly oily applications. Glove is designed to mold to the hand for improved movement and dexterity.

  • Ergonomic, light, flexible, and tough
  • Porous nitrile coating helps channel oil away from the glove's surface for a firm grip
  • Designed to mold to the hand for improved movement and dexterity
  • Ideal for quick and accurate handling
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Coated Area Palm, Fingers
Construction Cut and Sewn
Coating Material Nitrile Foam
Color Natural
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Cotton
Product Type Dip-coated Gloves
Size 10
For Use With (Application) Automotive, Machinery/Equipment, and Metal Fabrication
Glove Cuff Style Knit
Series 40-400
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.