Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell ActivArmr™ 27-600 Nitrile Coated Heavy Duty Work Gloves - Safety Cuff

Catalog No. 11391128B Shop All Ansell Products
Click to view available options
Size:
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves feature a unique coating formulation that provides increased resistance against sharp and abrasive environments, in addition to oil and grease.

  • Designed to be extra tough for use in environments that involve rough, abrasive materials
  • Constructed of cotton and jersey fabric for a high level of user comfort
  • Unique nitrile coating provides barrier protection against oil and grease
  • Safety cuff increases protection to the wrist
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Construction Cut and Sewn
Coating Material Nitrile
Color Blue, Natural
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Cotton
Product Type Dip-coated Gloves
Size 10
EN388 Standard 4121B
For Use With (Application) Automotive Assembly, Machinery/Equipment Operation, and Metal Fabrication
Glove Cuff Style Safety
Series ActivArmr™ 27-607
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.