Learn More
Ansell HyFlex™ Cut-Resistant Gloves
Catalog No. p-3565902 Shop All Ansell Products
Click to view available options
Size:
6
9
10
11
Hem Color:
Black
Blue
Brown
Purple
This item is not returnable. View return policy
Catalog No. 19-168-460
Supplier: Ansell 288734
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Offer a combination of comfort, softness, coolness and cut protection
  • Constructed of DSM Dyneema™, Spandex™ and nylon
  • Ergnomic design promotes wearer comfort
  • Proprietary black polyurethane coating provides sure grip
  • Can be washed and reused without any loss of cut protection
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Polyurethane
Color Black, White
Cut Resistance Rating ANSI A2
Material Dyneema™, Lycra
Product Type Cut-resistant Gloves
Size 6
For Use With (Application) Glass manufacturing, assembly of white goods, Plastics injection and moulding, automotive, assembling Metal parts and components, handling of sharp edged objects, preassembling, cutting of small dry or lightly Oiled parts and maintenance
Glove Cuff Style Knit
Hem Color Purple
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.