Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell HyFlex™ 11-900 Nitrile Coated Oil Repellent Gloves

Catalog No. 19148348 Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Industrial work gloves designed for light mechanical protection, particularly in moderately oily manufacturing or maintenance environments.

  • Nitrile coating provides light mechanical protection and dexterity
  • Designed for use in moderately oily manufacturing and maintenance environments
  • Combines protection with enhanced grip and abrasion resistance
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Chlorinated No
Coated Area Palm, Fingers
Construction Knit
Coating Material Nitrile
Color White, Blue
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size 9
For Use With (Application) Assembly, Equipment Operation, and Metal Fabrication
Gender Unisex
Glove Cuff Style Knit
Grip Design Rough
Series 11-900
Yarn Gauge 13
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.