Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell ActivArmr™ 47-400 Medium Duty Nitrile Coated Oil Repellent Gloves

Catalog No. 19019865 Shop All Ansell Products
Click to view available options
Size:
8
9
10
This item is not returnable. View return policy

Catalog No. 19-019865

Supplier: Ansell 205933

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

ActivArmr™ gloves combine the comfort of cotton or light leather gloves with the abrasion resistance and oil repellence of nitrile coated gloves.

  • ActivArmr™ gloves feature interlock knit lining, generous sizing, and contoured designs for improved comfort
  • Special nitrile formulation provides a combination of flexibility, mechanical resistance, and oil repellency
  • Natural hand shape is easier to wear for long periods
  • Gloves ensure cleanliness of metal sheets in the steel and motor industries
  • Silicone-free
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Construction Cut and Sewn
Coating Material Nitrile
Color Blue, Natural
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Cotton
Product Type Dip-coated Gloves
Size 9
For Use With (Application) Automotive, Machinery/Equipment, and Metal Fabrication
Glove Cuff Style Knit
Series 47-400
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.