Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell ActivArmr™ 28-359 Nitrile Coated Heavyweight Gloves with NitraGold™ Cuff

Catalog No. p-4591625 Shop All Ansell Products
Click to view available options
Size:
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves combine the strength of aramid fibers with the grip and flexibility of Ansell's advanced nitrile coating technology. NitraGold&Trade; cuff provides mechanical protection against cuts and abrasion.

  • Heavyweight design provides a substantial level of protection
  • Aramid fiber construction is strong and durable
  • Nitrile coating is oil resistant and enhances grip capabilities
  • NitraGold™ cuff provides mechanical protection against cuts and abrasion
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category III
Cuff Material NitraGold™
Coating Material Nitrile
Color Black, Gold
Cut Resistance Rating ANSI A3
Hand Specific
Product Type Dip-coated Gloves
Size 9
EN388 Standard 4321C
For Use With (Application) Automotive Assembly/Repair
Liner Material Aramid Fiber
Series ActivArmr™ 28-359
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.