Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell HyFlex™ 74-301 Lightweight Cut Protection Gloves

Catalog No. p-141986 Shop All Ansell Products
Click to view available options
Size:
8
9
Glove Cuff Color:
Blue
Green
This item is not returnable. View return policy
This item is not returnable. View return policy

Lightweight gloves provide comfort and dexterity for cut protection in food processing applications.

  • Ideal for use in meatpacking, food processing, materials handling and numerous other applications requiring a high level of protection from cuts
  • Full fingered, White, Ambidextrous
  • Tough triple-steel core is wrapped with long-lasting manmade fibers
  • Made from custom-spun yarns that contain three strands of stainless steel
  • Knitted construction contains no seams to irritate hands
  • Machine launderable, bleaching and hot-air drying will not affect performance
  • Color-coded wrist trim makes size identification quick and easy
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) CE, EN 420:2003 + A1:2009; Category II
Construction Knit
Color White
Cut Resistance Rating ANSI A8
Hand Ambidextrous
Latex Free Yes
Material Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size 8
EN388 Standard EN 388:1X4XF, EN 388:254X
For Use With (Application) Cleaning deli slicers, cutting up food, meat packing
Glove Cuff Color Green
Glove Cuff Style Knit
Liner Material Polyester, Stainless Steel
Model 74-301 8 222062
Series HyFlex™ 74-301
Length (English) 9 in.
Length (Metric) 22 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.