Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell HyFlex™ 48-101 Polyurethane Coated Seamless Knit Nylon Gloves

Catalog No. 18999455f Shop All Ansell Products
Click to view available options
Size:
6
8
9
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves are designed for light applications that require precise handling of small objects in dry or slightly oily environments.

  • Designed for light duty applications that require comfort and abrasion resistance
  • Polyurethane coating enhances grip in dry or light oil applications
  • Seamless construction prevents irritation
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Chlorinated No
Coated Area Palm
Construction Knit
Coating Material Polyurethane
Color Black
Cut Resistance Rating ANSI A1
Exterior Finish Rough
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size 11
For Use With (Application) Assembly, Inspection, and Packaging
Gender Unisex
Glove Cuff Style Knit
Grip Design Rough
Series 48-101
Yarn Gauge 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.