Learn More
Ansell AlphaTec™ 04-412 PVC-Nitrile Blend Coated Gauntlet Gloves
Catalog No. p-139016 Shop All Ansell Products
Click to view available options
Size:
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Combination PVC and nitrile gloves feature a secure wet grip while providing chemical, cut and abrasion resistance.
Ideal for applications involving petrochemicals and for handling equipment and abrasive materials coated with acids, caustics, solvents, grease, and oil.
  • Feature PVC-nitrile blend rough finish
  • Curved, preflexed fingers and wing thumb give extra comfort and flexibility
  • Two-piece lining eliminates seams from work areas to prevent irritation and premature wear-through
TRUSTED_SUSTAINABILITY
Specifications
Color Green
Latex Free Yes
Material PVC, Nitrile
Size Large
For Use With (Application) Chemical processing, oil refining and petrochemicals, cleaning and de-greasing with soaps and detergent formulations, handling equipment and abrasive materials coated with oil, grease, acids and solvents.
Glove Cuff Style Gauntlet
Liner Material Jersey Knit
Lining Lined
Model 04-412 9 211134
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.