Learn More
Ansell Textured Latex Sterile Gloves
Catalog No. p-3752737 Shop All Ansell Products
Change view
Click to view available options
Size:
6.5
7.5
8.5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-304989 8.5
19-304986 6.5
19-299951 7.5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-304989 Supplier Ansell Supplier No. 8607
This item is not returnable. View return policy
Sterile latex gloves with a textured finish for reliable grip. Very strong and durable construction with excellent tensile strength. Excellent comfort and fit due to its high elasticity and memory.
NO POWDER™ surgical gloves are made from latex and are powder free. The gloves have a straight, reinforced cuff for extra coverage and protection on the forearm, and a textured finish for superior grip and handling of instruments and sutures.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO9001, ISO13485, ASTM D 3577, Type 1
Color Cream
Exterior Finish Fully Textured
Material Latex
Powdered Powder Free
Size 8.5
Sterility Sterile
Thickness (English) 8.7 mil
Length (English) 11.8 in.
Length (Metric) 30 cm