Learn More
Ansell™ AlphaTec™ 56-910 Vinyl Closed-Back Aprons With Snap-on Ties
Catalog No. p-20830 Shop All Ansell Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Catalog No. 17-987-116A
Supplier: Ansell™ 950190
Only null left
Add to Cart
Add to Cart
With heat-sealed elastic cuffs
  • Neck ties, 15 in. long (38cm); waist ties, 29 in. long (74cm)
  • 8mil thick aprons have electronically welded vinyl binding strip that covers all edges for smoother finish
  • 6mil thick aprons have electronically welded binding strip around neck edge only; other edges are unhemmed
  • Available with snap-on nylon ties
  • Blue
Order Info
Can be cleaned to ISO Class 5 (Class 100) cleanroom standards and shipped in heat-sealed bags; call your Fisher Safety Customer Service Representative at 1-800-772-6733 for details.
Compliance
Permeation tested in accordance with MIL-STD 282 (using various chemical warfare agents) and ASTM™ F739 (using ASTM F1001 test battery).
TRUSTED_SUSTAINABILITY
Specifications
Size Small
Material Vinyl
Color Blue
Cuff Style Elastic
Thickness 8 mil
Includes Snap-on ties
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.