Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anthrax EF Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB222538 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB222538 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB222538 Supplier Novus Biologicals Supplier No. NBP241047B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anthrax EF Polyclonal antibody specifically detects Anthrax EF in Bacteria samples. It is validated for ELISA

Specifications

Antigen Anthrax EF
Applications ELISA
Classification Polyclonal
Conjugate Biotin
Dilution ELISA
Formulation PBS
Gene Alias Adenylyl cyclase, Anthrax edema toxin adenylate cyclase component, ATP pyrophosphate-lyase, Calmodulin-sensitive adenylate cyclase, Edema factor 1 (EF 1)
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to 16 amino acids near the carboxy terminus of the Anthrax edema factor protein. The immunogen is located within the last 50 amino acids of Anthrax Edema Factor. Amino Acid Squence: NFTENETDNFEVFQK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.