Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anthrax LF Antibody (5F502.2), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB339507 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339507 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339507 Supplier Novus Biologicals Supplier No. NB10056592JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anthrax LF Monoclonal specifically detects Anthrax LF in Human samples. It is validated for Western Blot.

Specifications

Antigen Anthrax LF
Applications Western Blot
Classification Monoclonal
Clone 5F502.2
Conjugate Janelia Fluor 646
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Anthrax lethal factor, Anthrax lethal toxin endopeptidase component, Anthrax LF, B.anthracis lethal factor, bacillus anthracis lethal factor, Lef, LF
Host Species Mouse
Immunogen The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to amino acids 779-792 of the Anthrax LF protein (approx. 90 kD).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.