Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anthrax PA Antibody, Novus Biologicals™
SDP

Catalog No. NB10056670 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056670 0.2 mL
NB10056670T 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056670 Supplier Novus Biologicals Supplier No. NB10056670
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anthrax PA Polyclonal specifically detects Anthrax PA in Bacteria samples. It is validated for Western Blot.

Specifications

Antigen Anthrax PA
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml to 10 ug/ml
Formulation Whole Rabbit antisera diluted with PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias anthrax PA, Anthrax toxins translocating protein, Bacillus anthracis protective antigen, PA, PA 83, PA83, PAG, Paga, Protective antigen
Host Species Rabbit
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 186-192 of the Anthrax PA protein (approx. 83 kDa).
Purification Method Unpurified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity PA is cleaved from the 83 kDa form to the 63 kDa form at amino acids 164-167. Thus the antibody (made against amino acids 186-192) should recognize both the 83 kDa and 63 kDa PA forms, but not the 20 kDa cleavage fragment.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.