Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anthrax PA Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $366.00

Specifications

Antigen Anthrax PA
Dilution Western Blot 2 ug/ml to 10 ug/ml
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056670T
SDP
View Documents
Novus Biologicals
NB10056670SS
0.05 mL
Each for $181.90
Only null left
Add to Cart
 
NB10056670
SDP
View Documents
Novus Biologicals
NB10056670
0.2 mL
Each for $366.00
Only null left
Add to Cart
 
Description

Description

Anthrax PA Polyclonal specifically detects Anthrax PA in Bacteria samples. It is validated for Western Blot.
Specifications

Specifications

Anthrax PA
Western Blot
Unconjugated
Rabbit
Bacteria
anthrax PA, Anthrax toxins translocating protein, Bacillus anthracis protective antigen, PA, PA 83, PA83, PAG, Paga, Protective antigen
IgG
Unpurified
PA is cleaved from the 83 kDa form to the 63 kDa form at amino acids 164-167. Thus the antibody (made against amino acids 186-192) should recognize both the 83 kDa and 63 kDa PA forms, but not the 20 kDa cleavage fragment.
Western Blot 2 ug/ml to 10 ug/ml
Polyclonal
Antisera
RUO
Whole Rabbit antisera diluted with PBS and 0.05% BSA with 0.05% Sodium Azide
The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 186-192 of the Anthrax PA protein (approx. 83 kDa).
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.