Learn More
Description
Specifications
Specifications
| Antigen | Anthrax PA |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000 |
| Formulation | Whole antisera with 0.2 ml sterile PBS with No Preservative |
| Gene Alias | anthrax PA, Anthrax toxins translocating protein, Bacillus anthracis protective antigen, PA, PA 83, PA83, PAG, Paga, Protective antigen |
| Host Species | Rabbit |
| Immunogen | The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 186-192 of the Anthrax PA protein (approx. 83 kDa). |
| Purification Method | Unpurified |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
