Learn More
Anthrone, ACS
Description
Used for determination of carbohydrates Anthrone is used for the determination of carbohydrates. It is used as a general reagent in the colorimetric determination of carbohydrates in yeast and plant extracts. It is involved in organic synthesis. It is utilized for the determination of sugar in body fluids. It finds application as a laxative.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 155°C to 158°C |
| Quantity | 25 g |
| Beilstein | 1910173 |
| Sensitivity | Light sensitive |
| Merck Index | 14,691 |
| Solubility Information | Soluble in acetone,hot benzene,concentrated sulfuric acid,hot dilute alkali and most organic solvents. Insoluble in water. |
| Formula Weight | 194.23 |
| Grade | ACS |
| Physical Form | Crystalline |
| Chemical Name or Material | Anthrone |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 201-994-0 |
| RTECSNumber | CB8925500 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.