Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HTR3E Rabbit anti-Human, Polyclonal, Invitrogen™

Catalog No. PIPA524029
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524029 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524029 Supplier Thermo Scientific Supplier No. PA524029

Rabbit Polyclonal Antibody

The product of this gene belongs to the ligand-gated ion channel receptor superfamily. HTR3E encodes a subunit E of the type 3 receptor for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor causes fast, depolarizing responses in neurons after activation. Genes encoding subunits C, D and E form a cluster on chromosome 3. An alternative splice variant has been described but its full length sequence has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HTR3E
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene HTR3E
Gene Accession No. A5X5Y0
Gene Alias 5-HT3-E, 5-HT3E, 5-HT3c1, 5-hydroxytryptamine (serotonin) receptor 3, family member E, 5-hydroxytryptamine receptor 3 subunit E, 5-hydroxytryptamine receptor 3E
Gene Symbols HTR3E
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 126-154 amino acids from the central region of human 5HT3E
Purification Method Antigen affinity chromatography
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285242
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.