Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ACK1, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP132311 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP132311 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP132311 Supplier Novus Biologicals Supplier No. NBP132311

Rabbit Polyclonal Antibody

ACK1 Polyclonal specifically detects ACK1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ACK1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Alias ACK-1, ACK1ACKp21cdc42Hs, activated CDC42 kinase 1, activated Cdc42-associated kinase 1, activated p21cdc42Hs kinase, EC 2.7.10, EC 2.7.10.2, FLJ44758, FLJ45547, Tyrosine kinase non-receptor protein 2, tyrosine kinase, non-receptor, 2
Gene Symbols TNK2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human ACK1. The exact sequence is proprietary.
Molecular Weight of Antigen 115 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10188
Test Specificity Tested in human. Not yet tested in any other species.
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.