Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Acrolein, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB200556 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB200556 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB200556 Supplier Novus Biologicals Supplier No. NB200556

Specifications

Antigen Acrolein
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, ELISA 1:5000-1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:500-1:1000, Dot Blot
Formulation PBS (pH 7.2) with No Preservative
Gene Alias Acrylic amide; Ethylene Carboxamide 2-Propenamide; 2-Propen-1-one; 2-propenal; Acraldehyde; Acrolein; Acrylic aldehyde
Host Species Rabbit
Immunogen Synthetic acrolein conjugated to protein carrier (PC).
Purification Method Immunogen affinity purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes conjugated Acrolein. Using a conjugate Acrolein Protein, specificity was performed with an ELISA test by competition experiments with the following compounds. Acrolein-BSA (ratio 1:1), Malondialdehyde-BSA (1:55), Glyoxal-BSA (1:>50,000), Formaldehyde-BSA (1:>50,000), Glutaraldehyde-BSA (1:> 50,000)
Reconstitution Reconstitute with 0.1 ml sterilized deionized water.
Target Species Non-species specific
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Precipitation
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.