Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Skeletal Muscle Actin, Polyclonal, Invitrogen™

Catalog No. 01673020
Change view
Click to view available options
Quantity:
7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-673-020 7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-673-020 Supplier Thermo Scientific Supplier No. PA137019

Rabbit Polyclonal Antibody

This is highly specific and shows no cross-reaction with smooth muscle actin. This antibody reacts with sarcomeric actins of normal tissues and neoplasms derived from such tissues (i.e. rhabdomyosarcomas).

This antibody is predicted to react with bovine, rabbit, sheep, rat, guinea pig, and frog based on sequence homology.A suggested positive control is skeletal muscle tissue.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Skeletal Muscle Actin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Gene ACTA1
Gene Accession No. P68133
Gene Alias ACTA1
Gene Symbols ACTA1
Host Species Rabbit
Immunogen Synthetic peptide derived from N-terminus of human alpha skeletal muscle isoform of actin
Purification Method Antigen affinity chromatography
Quantity 7 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58
Target Species Human
Content And Storage Store at 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.