Learn More
Adaptin α Mouse, Unlabeled, Clone: 8, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 610501
Description
Sorting of integral membrane proteins at various stages of the endocytic and secretory pathways is mediated by vesicular trafficking between a variety of organelles. Two sorting signals are tyrosine-based and dileucine-based signals that interact with heterotetrameric adaptor protein complexes (AP-1, AP-2, AP-3, and AP-4), which are associated with the vesicle coats. These coatomers contain two large Adaptin proteins (α, γ, δ, or ε and β1, β2, β3 or β4, respectively) that are noncovalently linked to one medium chain (μ1, μ2, μ3 or μ4) and one small chain (σ1, σ2, σ3 or σ4). The AP-1 and AP-3 complexes are involved in protein sorting from the TGN and endosomes, while AP-2 adaptor complexes are involved in clathrin-mediated endocytosis. In the AP-2 complex, Adaptin α is expressed in two very similar isoforms from two different genes. Adaptin α [A] (112kDa) is expressed primarily in brain, while Adaptin α [C] (105kDa) is expressed in brain, liver and other tissues.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications
Adaptin α | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Mouse Adaptin α [A] aa. 38-215 | |
50 μg | |
Cell Biology | |
Canine, Human, Mouse, Rat | |
IgG1 |
Western Blot | |
8 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.