Learn More
AFAP Mouse, Unlabeled, Clone: 36, BD
Mouse Monoclonal Antibody
Specifications
Antigen | AFAP |
---|---|
Clone | 36 |
Concentration | 250μg/mL |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Chicken embryo fibroblasts (CEF) which are transformed by pp60[src] show anincrease in the steady-state levels of tyrosine-phosphorylated proteins. One of these proteins is the Actin Filament-Associated Protein, AFAP110. This protein forms a stable complex with activated pp60[src] via the SH3 and SH2 domains ofthe src protein. However, tyrosine phosphorylation of AFAP requires only the intact src SH3 domain. AFAP is associated with actin stress fibers in normal CE cells, but following transformation by pp60[src], AFAP becomes localized in rosette-like structures known as podosomes. Several sequence motifs within AFAP110 are predicted to bind to SH2 and to SH3 domains. In vitro analysis of AFAP in normal cell extracts demonstrated that it binds less efficiently to Src SH2-containing fusion proteins than does AFAP from src-transformed cell extracts.
Specifications
AFAP | |
250μg/mL | |
Unconjugated | |
RUO | |
Chicken, Human, Mouse, Rat | |
IgG1 | |
Affinity Purified |
36 | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.