Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIP1 Mouse, Unlabeled, Clone: 49, BD

Catalog No. p-3850320 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611620 50 μg
BDB611621 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611620 Supplier BD Biosciences Supplier No. 611620
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Apoptosis is a selective process of genetically programmed cell death which occurs during normal cell differentiation and development of multicellular organisms. In vertebrates, T cell and neuronal development are probably the best characterized systems for the study of apoptosis. ALG-2 and ALG-3 (apoptosis-linked genes 2 and 3) were identified as low molecular weight Ca2+ -binding proteins essential for apoptosis through the activation of the Fas receptor in T cells. ALG-2 Interacting Protein 1 (AIP1/Alix) is a ubiquitous protein that associates with ALG-2 in the cytosol in a Ca2+; dependent manner. AIP1 is homologous to the yeast protein, BRO1, which has been implicated in Pkc1p- AP kinase signaling. A truncated form of AIP1 protects against serum starvation-, etoposide-, and staurosporine-induced cell death. In addition, the C-terminal proline rich region of AIP1 facilitates interaction with SH3 domain-containing protein expressed in tumorigenic astrocytes (SETA) and this interaction may be important for mediating DNA damage-dependent apoptosis in astrocytes. Thus, AIP1 interacts with ALG-2 or SETA, or both, during activation of cell death pathways in a variety of cell types.

Host Species: Mouse
Clone: 49
Isotype: IgG1
Species Reactivity [for Features Main]: Rat
Immunogen: Mouse AIP1 aa. 375-580

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen AIP1
Applications Western Blot
Classification Monoclonal
Clone 49
Concentration 250μg/mL
Conjugate Unconjugated
Description ALG-2 Interacting Protein 1; Alix
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse AIP1 aa. 375-580
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.