Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH Mouse anti-Human, Unlabeled, Clone: 44, BD

Catalog No. p-3968377 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611194 50 μg
BDB611195 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611194 Supplier BD Biosciences Supplier No. 611194
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Aldehyde dehydrogenase (ALDH) is a ubiquitous enzyme located in nearly all mammalian tissues. It catalyzes the irreversible oxidation of a range of aliphatic and aromatic aldehydes to their corresponding carboxylic acids. There are multiple isoforms of ALDH which are subdivided into three classes. Class I includes the cytosolic isoforms. Class II includes the mitochondrial isoforms. Class III includes the microsomal, cytosolic tumor specific, and cytosolic dioxin-inducible forms. At least twelve human ALDH isoforms have been identified. Mutations of many of these proteins such as ALDH1, ALDH2, ALDH4, and ALDH10 have been implicated in multiple human metabolic disorders and clinical abnormalities. At the amino acid level, human ALDH isoforms exhibit a wide range of diversity (15% to about 80%). However, multiple protein regions have been highly conserved and are important for functional activities. A well-characterized member of the human ALDH family is ALDH1. It plays a major role in the biosynthesis of retinoic acid from retinol (vitamin A). Retinoic acid, the biologically
active form of retinol, is a regulator of cellular proliferation, differentiation, and survival.

Host Species: Mouse
Clone: 44
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human ALDH1 aa. 7-128
Formula Weight [Chemical]: 55kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH
Applications Western Blot
Classification Monoclonal
Clone 44
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human ALDH1 aa. 7-128
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.