Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

aldehyde dehydrogenase 1 family, member L1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004516 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-516 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-516 Supplier Abnova Corporation Supplier No. H00010840A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant ALDH1L1.

The protein encoded by this gene catalyzes the conversion of 10-formyltetrahydrofolate, NADP, and water to tetrahydrofolate, NADPH, and carbon dioxide. The encoded protein belongs to the aldehyde dehydrogenase family and is responsible for formate oxidation in vivo. Deficiencies in this gene can result in an accumulation of formate and subsequent methanol poisoning. [provided by RefSeq

Sequence: EESFGPVMIISRFADGDLDAVLSRANATEFGLASGVFTRDINKALYVSDKLQAGTVFVNTYNKTDVAAPFGGFKQSGFGKDLGEAALNEYLRVKTVTFEY

Specifications

Antigen aldehyde dehydrogenase 1 family, member L1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ALDH1L1.
Formulation 50% glycerol
Gene ALDH1L1
Gene Accession No. NM_012190
Gene Alias DKFZp781N0997/FTHFD
Gene Symbols ALDH1L1
Host Species Mouse
Immunogen ALDH1L1 (NP_036322, 803 a.a. ∼ 902 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 10840
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.