Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin VII Mouse anti-Human, Unlabeled, Clone: 5, BD

Catalog No. BDB610669 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610669 150 μg
BDB610668 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610669 Supplier BD Biosciences Supplier No. 610669

Mouse Monoclonal Antibody

Annexin VII, also known as synexin, is a member of the Annexin family characterized by Ca2+ dependent phospholipid binding. Annexins have a four-fold internal repeat of about 70 amino acids that contains the Ca2+-binding sites. They also have a regulatory NH2-terminal region of 30-40 amino acids. Annexin VII mRNA has been detected in many tissues and is most abundant in brain, heart, skeletal muscle, and lung. Annexin VII mRNA exhibits a tissue dependent polymorphism due to alternative splicing which results in two different isoforms of 47kDa and 51kDa. In vitro, Annexin VII aggregates chromaffin granules and enhances membrane fusion in a Ca2+ and GTP-dependent manner. Although the physiological role of Annexin VII is not completely understood, it is believed to influence and regulate Ca2+ dependent events such as secretion at the plasma membrane.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Annexin VII
Applications Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 5
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.