Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ApoA-1 Mouse anti-Rat, Unlabeled, Clone: 4, BD

Catalog No. BDB612330 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612330 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612330 Supplier BD Biosciences Supplier No. 612330

Mouse Monoclonal Antibody

In blood, cholesterol and triglycerides are transported in lipoprotein particles that consist of a single layer of phospholipid surrounding a lipid core and surface-associated apolipoproteins (Apo). The Apo-proteins are involved in the specific binding of cellular receptors, the regulation of lipolytic enzymes, and the process of lipid exchange. High density lipoprotein particles (HDLs) contain the apolipoproteins ApoA-I, ApoA-II and Apo-M, whereas low density (LDL), intermediate density, and very low density (VLDL) lipoprotein particles contain ApoB-100 as the primary structural element. ApoA-I is synthesized in the liver and small intestines, where it acts as a cofactor for lecithin-cholesterol acyltransferase during the formation of cholesterol ester. ApoA-II is synthesized in the liver where it activates hepatic lipase. In HepG2 cells, treatment with gramoxone causes oxidative stress and reductions in ApoA-I mRNA levels. This down regulation of ApoA-I may contribute to reduced plasma HDL levels in response to oxidants, such as cigarette smoke. Thus, ApoA-I is an important cofactor fo cholesterol synthesis and is a major component of HDLs.

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen ApoA-1
Applications Western Blot
Classification Monoclonal
Clone 4
Concentration 250μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Rat ApoA-I aa. 144-258
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Rat
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.