Learn More
ApoA-1 Mouse anti-Rat, Unlabeled, Clone: 4, BD
Mouse Monoclonal Antibody
Specifications
| Antigen | ApoA-1 |
|---|---|
| Clone | 4 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
BDB612330
|
BD Biosciences
612330 |
50 μg | N/A | N/A | N/A | ||||
Description
In blood, cholesterol and triglycerides are transported in lipoprotein particles that consist of a single layer of phospholipid surrounding a lipid core and surface-associated apolipoproteins (Apo). The Apo-proteins are involved in the specific binding of cellular receptors, the regulation of lipolytic enzymes, and the process of lipid exchange. High density lipoprotein particles (HDLs) contain the apolipoproteins ApoA-I, ApoA-II and Apo-M, whereas low density (LDL), intermediate density, and very low density (VLDL) lipoprotein particles contain ApoB-100 as the primary structural element. ApoA-I is synthesized in the liver and small intestines, where it acts as a cofactor for lecithin-cholesterol acyltransferase during the formation of cholesterol ester. ApoA-II is synthesized in the liver where it activates hepatic lipase. In HepG2 cells, treatment with gramoxone causes oxidative stress and reductions in ApoA-I mRNA levels. This down regulation of ApoA-I may contribute to reduced plasma HDL levels in response to oxidants, such as cigarette smoke. Thus, ApoA-I is an important cofactor fo cholesterol synthesis and is a major component of HDLs.
Western Blotting
Specifications
| ApoA-1 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Rat ApoA-I aa. 144-258 | |
| Primary |
| 4 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Rat | |
| IgG1 | |
| Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.