Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Arrestin Mouse, Unlabeled, Clone: 10, BD

Catalog No. p-3856065 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610550 50 μg
BDB610551 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610550 Supplier BD Biosciences Supplier No. 610550
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

β-Arrestins were discovered due to their ability to modulate interactions between the phosphorylated β2-Adrenergic receptors and G proteins. This modulation results in diminished β2-Adrenergic receptor function, also known as desensitization. Because arrestins are found at the synaptic terminals, they may provide a termination mechanism that allows the neurons to regain their original polarization and respond to a new neurotransmitter stimulus. The C-terminal region of arrestins is involved in selecting the phosphorylated and activated adrenergic receptors. The β-Arrestin1 gene encodes a protein of 418 amino acids with an approximate molecular weight of 55kDa. β-Arrestin1 protein is highly homologous to the 45kDa β-Arrestin2. Both proteins are widely expressed, but are especially abundant in the central nervous system.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen β-Arrestin
Applications Western Blot
Classification Monoclonal
Clone 10
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat β-Arrestin1 aa. 262-409
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.