Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASF-1 (SF2) Mouse anti-Human, Clone: AK96, MilliporeSigma™

Catalog No. MABE163MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE163MI 1 Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE163MI Supplier MilliporeSigma Supplier No. MABE163
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-ASF-1 (SF2) Antibody, clone AK96 (mouse monoclonal antibody) validated in western blot, immunoprecipitation, immunocytochemistry, immunohistochemistry to detect ASF-1 (SF2) also known as splicing factor arginine/serine-rich 1, Alternative-splicing factor 1 and pre-mRNA-splicing factor SF2 P33 subunit
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASF-1 (SF2)
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone AK96
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl without 0.05% sodium azide.
Gene Accession No. NP_001071634
Gene Symbols SF2, ASF, ASF-1, SF2p33, SF2P33, SRp30a
Host Species Mouse
Immunogen MBP-tagged recombinant protein corresponding to the N-terminus of human ASF-1 (SF2).
Purification Method Protein G purified
Quantity 1 Unit
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.