Learn More
ATP Synthase α Mouse, Unlabeled, Clone: 51, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 612517
Description
ATP synthase is a large enzyme complex that uses an electrochemical H+ or Na+ gradient to synthesize ATP from ADP and Pi, providing the organism with the ATP needed for energy. The complex consists of two major units, F0 and F1. F0 is embedded in the inner membrane of the mitochondria and, due to its hydrophobic nature, translocates protons across this membrane. F1 is the catalytic portion in the matrix region of the mitochondria and is comprised of α, β, γ, δ, and ε subunits at a 3:3:1:1:1 ratio. The α subunit is a ubiquitous protein that is highly conserved among species. It has an adenine specific binding site that binds both ATP and ADP. There are two glycine rich regions in the sequence, the A domain and B domain, that are thought to be part of the nucleotide binding domain. It has been demonstrated that the N-terminus of the α subunit is necessary for the correct functional and structural connection of F0 to F1. Thus, the α subunit is an essential component of the ATP synthase complex and plays a role in properly orienting the F0 and F1 units.
Immunofluorescence, Western Blotting

Specifications
ATP Synthase α | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Human ATP Synthase α aa. 113-220 | |
150 μg | |
Cell Biology | |
Canine, Chicken, Human, Mouse, Rat | |
IgG2a |
Western Blot | |
51 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.