Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATPase, H+ transporting, lysosomal, V0 subunit d2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004822 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-822 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-822 Supplier Abnova Corporation Supplier No. H00245972A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant ATP6V0D2.

Sequence: ETLYPTFGKLYPEGLRLLAQAEDFDQMKNVADHYGVYKPLFEAVGGSGGKTLEDVFYEREVQMNVLAFN

Specifications

Antigen ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ATP6V0D2.
Formulation 50% glycerol
Gene ATP6V0D2
Gene Accession No. NM_152565
Gene Alias ATP6D2/FLJ38708/VMA6
Gene Symbols ATP6V0D2
Host Species Mouse
Immunogen ATP6V0D2 (NP_689778, 238 a.a. ∼ 306 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 245972
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.