Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl-X set Mouse, Unlabeled, Clone: 2H12, BD
Mouse Monoclonal Antibody
$444.00
Specifications
Antigen | Bcl-X set |
---|---|
Clone | 2H12 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Members of the Bcl-2 family play a major role in regulating the response of cells to a wide variety of apoptotic signals. Some Bcl-2 family members like Bcl-2, block apoptosis, whereas others promote apoptosis and inhibit Bcl-2 activity. Bcl-x is a Bcl-2 homologue that has two isoforms, resulting from alternative splicing. Bcl-xL (long) is a 241 amino acid protein (25-29 kDa) that is 47% homologous to Bcl-2 on the amino acid level. Bcl-xS (short) is a 178 amino acid protein (∼ 19.5 kDa) lacking a 63 amino acid domain that is well conserved among members of the Bcl-2 protein family. Bcl-xL blocks cell death, whereas Bcl-xS inhibits Bcl-2 and promotes cell death.Specifications
Bcl-X set | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse | |
Primary | |
Store undiluted at -20°C. |
2H12 | |
Monoclonal | |
Mouse | |
Cell Biology | |
IgG2a | |
Affinity Purified |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title