Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl-X Mouse, Unlabeled, Clone: 44, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 610747
Description
Bcl-x is related to the Bcl-2 protein and can function independently of Bcl-2 in regulating apoptosis (programmed cell death). The bcl-x transcript is expressed in a number of tissues, with the highest levels in the lymphoid and central nervous systems. Two distinct cDNA species, bcl-xL and bcl-xS, have been observed and appear to arise from alternate 5' splice sites located within the first coding exon of the bcl-x gene. Bcl-xL is composed of 233 amino acids and is similar in size and structure to Bcl-2. The Bcl-xS polypeptide shows a deletion of 63 amino acids which consist of the region of Bcl-xL with the highest degree of amino acid identity to Bcl-2. Like Bcl-2, Bcl-xL inhibits cell death upon growth factor withdrawal when transfected into an IL-3-dependent cell line. However, Bcl-xS inhibits Bcl-2 mediated cell survival.
Specifications
Bcl-X | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Human Bcl-Xl aa. 18-233 | |
150 μg | |
Cell Biology | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
44 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction