Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl-X Mouse, Unlabeled, Clone: 44, BD
Mouse Monoclonal Antibody
$280.00 - $556.00
Specifications
Antigen | Bcl-X |
---|---|
Clone | 44 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
Bcl-x is related to the Bcl-2 protein and can function independently of Bcl-2 in regulating apoptosis (programmed cell death). The bcl-x transcript is expressed in a number of tissues, with the highest levels in the lymphoid and central nervous systems. Two distinct cDNA species, bcl-xL and bcl-xS, have been observed and appear to arise from alternate 5' splice sites located within the first coding exon of the bcl-x gene. Bcl-xL is composed of 233 amino acids and is similar in size and structure to Bcl-2. The Bcl-xS polypeptide shows a deletion of 63 amino acids which consist of the region of Bcl-xL with the highest degree of amino acid identity to Bcl-2. Like Bcl-2, Bcl-xL inhibits cell death upon growth factor withdrawal when transfected into an IL-3-dependent cell line. However, Bcl-xS inhibits Bcl-2 mediated cell survival.Specifications
Bcl-X | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
44 | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
Human Bcl-Xl aa. 18-233 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title