Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Bone Sialoprotein Clone: ID1.2,

Catalog No. 203637100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
20-363-7100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-363-7100UG Supplier MilliporeSigma™ Supplier No. 203637100UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Bone Sialoprotein Clone: ID1.2 in Human samples, and it is validated for ELISA, Immunohistochemistry (Paraffin), Immuno Blotting

Except for osteocalcin and osteonectin, bone sialoprotein (BSP) is one of the major noncollagenous proteins in the extracellular matrix of bone. BSP could be useful as a potential serum marker for monitoring bone remodeling in diseases concerning bone turnover, pathological bone alterations and the rapid increase of osteoporosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bone Sialoprotein
Applications ELISA, Immunoblot, Immunoblot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ID1.2
Conjugate Unconjugated
Formulation Lyophilized from 50mM Tris, pH 7.4.
Host Species Mouse
Immunogen purified human bone sialoprotein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽70-80kDa bone sialoprotein in osteoblasts and osteocytes.
Target Species Human
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.