Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BrdU Mouse anti-Human, FITC, Clone: 3D4, BD

Catalog No. bdb556028 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556028 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556028 Supplier BD Biosciences Supplier No. 556028
Only null left
Add to Cart
Add to Cart

Includes

FITC Mouse Anti-BrdU (5-bromo-2'-deoxyuridine, 5-Bromouracil deoxyriboside, BUdR), FITC Mouse IgG1 κ Isotype Control

Mouse Monoclonal Antibody

Reacts with bromodeoxyuridine (BrdU) incorporated in place of thymidine during the S-phase of cycling cells. This technique enables measurement of the frequency of DNA-synthesizing cells in studies of cell activation and proliferation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen BrdU
Applications Flow Cytometry
Classification Monoclonal
Clone 3D4
Conjugate FITC
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.