Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-C1q, HRP, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB10062413 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062413 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062413 Supplier Novus Biologicals Supplier No. NB10062413

Specifications

Antigen C1q
Applications ELISA
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate HRP
Dilution ELISA 1:100-1:400
Formulation PBS and HRP stabilizer with 0.01% Thimerosal
Gene Alias A chain, complement component 1, q subcomponent, A chain, complement component 1, q subcomponent, alpha polypeptide
Gene Symbols C1QA
Host Species Sheep
Immunogen Human C1q, purified from plasma
Purification Method IgG purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 712
Test Specificity Recognizes human C1q, a sub-component of the C1 complement component.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.