Learn More
Calcineurin Mouse, Unlabeled, Clone: 29, BD
Mouse Monoclonal Antibody
$287.50
Specifications
Antigen | Calcineurin |
---|---|
Clone | 29 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
Calcineurin is a Ca2+/calmodulin-dependent protein phosphatase. The active enzyme is a heterodimer of a large calmodulin-binding catalytic subunit A (61kDa) and a smaller Ca2+ binding subunit B (19kDa). Regions corresponding to the calmodulin-binding site, an autoinhibitory domain, and a putative subunit B binding site have been identified within the large subunit A. The activity of calcineurin is sensitive to immunosuppressants such as cyclosporin A (CsA) and tacrolimus (FK506). The study of FK506-mediated inhibition of nitric oxide formation has revealed that nitric oxide synthase (NOS) is a calcineurin substrate. Calcineurin dephosphorylates NOS and enhances its catalytic activity. Therefore, Calcineurin is an essential mediator for efficient T cell antigen receptor (TCR)-mediated T cell activation.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
Calcineurin | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG2a | |
Affinity Purified |
29 | |
Western Blot | |
Unconjugated | |
RUO | |
Chicken, Frog, Human, Mouse, Rat | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.