Learn More
Calnexin Mouse, Unlabeled, Clone: 37, BD
Mouse Monoclonal Antibody
$281.50 - $556.00
Specifications
Antigen | Calnexin |
---|---|
Clone | 37 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
Calnexin is a 90kDa integral membrane protein located primarily in the ER. It contains a long N-terminal calcium-binding domain that extends into the lumen of the ER and a short, acidic cytosolic domain. Calnexin associates with several cell surface proteins as they pass through the ER. Since it also forms complexes with other resident ER proteins involved in the Ca2+-dependent retention of proteins, calnexin may act as a chaperone. The amino acid sequence of calnexin is highly conserved among species and is similar in sequence to calreticulin, another Ca2+-binding protein found in the ER. Neither calnexin nor calreticulin contain the calcium-binding motif, known as the “E-F hand” found in the calmodulin family of proteins.
Host Species: Mouse
Clone: 37
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human Calnein aa. 116-301
Immunofluorescence, Immunoprecipitation
Specifications
Calnexin | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
37 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Human, Mouse, Rat | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.