Learn More
CaM Kinase Kinase Mouse, Unlabeled, Clone: 6, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 610545
Description
Ca2+/Calmodulin (CaM)-dependent kinases are multifunctional kinases involved in a myriad of cellular functions such as neurotransmitter synthesis, long-term potentiation, and formation of spatial learning. A 68kDa CaM Kinase Kinase with homology to the Ca2+/CaM-dependent kinases, phosphorylates and activates Ca2+/CaM-dependent protein kinases I and IV, but not type II. This phosphorylation occurs on Ser/Thr residues and is Ca2+ and CaM dependent. CaM Kinase Kinase is abundant in brain tissue, and is thought to be an upstream regulator in the Ca2+/CaM-dependent neural processes.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications
CaM Kinase Kinase | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
150 μg | |
Cell Biology | |
Canine, Human, Rat | |
IgG2a |
Western Blot | |
6 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.