Learn More
CaM Kinase Kinase Mouse, Unlabeled, Clone: 6, BD
Mouse Monoclonal Antibody
$336.00 - $627.50
Specifications
| Antigen | CaM Kinase Kinase |
|---|---|
| Clone | 6 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Ca2+/Calmodulin (CaM)-dependent kinases are multifunctional kinases involved in a myriad of cellular functions such as neurotransmitter synthesis, long-term potentiation, and formation of spatial learning. A 68kDa CaM Kinase Kinase with homology to the Ca2+/CaM-dependent kinases, phosphorylates and activates Ca2+/CaM-dependent protein kinases I and IV, but not type II. This phosphorylation occurs on Ser/Thr residues and is Ca2+ and CaM dependent. CaM Kinase Kinase is abundant in brain tissue, and is thought to be an upstream regulator in the Ca2+/CaM-dependent neural processes.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
| CaM Kinase Kinase | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG2a | |
| Affinity Purified |
| 6 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Canine, Human, Rat | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.