Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cardiac Troponin T Mouse, PE, Clone: 13-11, BD

Catalog No. BDB564767 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564767 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564767 Supplier BD Biosciences Supplier No. 564767
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 13-11 monoclonal antibody specifically recognizes Troponin T type 2 (cardiac), encoded by the gene TNNT2 . Troponin T is the tropomyosin-binding subunit of the troponin complex, which also encompasses troponin C and troponin I. This complex regulates muscle contraction in skeletal and cardiac muscle in response to alterations in calcium levels. Troponin T type 2 is solely found in the heart, and genetic alterations in the TNNT2 gene are associated to a series of heart disorders in humans, including hypertrophic cardiomyopathy, dilated cardiomyopathy and left ventricular noncompaction. Cardiac Troponin T can be used as a marker for the identification of cardiomyocytes derived from pluripotent stem cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cardiac Troponin T
Applications Flow Cytometry
Classification Monoclonal
Clone 13-11
Conjugate PE
Description cTnT, Cardiac Muscle Troponin T, Troponin T type 2, TNNT2
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rabbit cardiac troponin T Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Canine, Chicken, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.