Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-3/CPP32 Mouse anti-Mouse, Unlabeled, Clone: 46, BD

Catalog No. BDB611049 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611049 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611049 Supplier BD Biosciences Supplier No. 611049
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Apoptosis, a selective process of genetically programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptotic cells are characterized by loss of cell volume, plasma membrane blebbing, nuclear condensation, chromatin aggregation, and endonucleocytic degradation of DNA into nucleosomal fragments. Caspase-3 (CPP32, Yama, apopain) is a member of the family of cysteine proteases which includes interleukin-1beta-converting enzyme (ICE) and C. elegans protein, Ced-3. An apoptotic signal such as granzyme B of cytotoxic T-cells (CTLs) or ICE-like proteases induces the intracellular cleavage of Caspase-3 from the inactive proform (32kDa) to the active form which consists of the p20, p17, and p12 subunits. The active form of Caspase-3 cleaves several other apoptotic proteins including proteins such as DNA fragmentation factor (DFF). Apoptosis can be inhibited by coexpression of Bcl-2 as well as inhibitors of Caspase-3 or other members of the family of cysteine proteases. This antibody recognizes the mouse 32kDa pro-Caspase-3 and the p17 cleaved form of Caspase 3 in T cell lymphocytes treated with camptothecin.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase-3/CPP32
Applications Western Blot
Classification Monoclonal
Clone 46
Concentration 250μg/mL
Conjugate Unconjugated
Description CPP32
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse CPP32 aa. 25-145
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.