Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CCR3, Alexa Fluor 700, Clone: 83101, R&D Systems™
Supplier: R&D Systems FAB729N025
This item is not returnable.
View return policy
Description
Host Species: Rat
Clone: 83101
Isotype: IgG2A
Species Reactivity: Ms
Immunogen: Y3 rat myeloid cell line transfected with mouse CCR3. Met1-Phe359
Applications:
Flow Cytometry
Specifications
CCR3 | |
Monoclonal | |
Alexa Fluor 700 | |
P51678 | |
CCR3 | |
Y3 rat myeloid cell line transfected with mouse CCR3, Met1-Phe359, Accession # P51678 | |
25 Tests | |
1232 | |
Mouse | |
Purified |
Flow Cytometry | |
83101 | |
Flow Cytometry 5 uL/10^6 cells | |
b-chemokine receptor, CC chemokine receptor 3, C-C chemokine receptor type 3, C-C CKR-3, CCR3, CCR-3, CD193, CD193 antigen, chemokine (C-C motif) receptor 3, CKR3CC-CKR-3CMKBR3CD193, eosinophil CC chemokine receptor 3, Eosinophil eotaxin receptor, MGC102841 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects mouse CCR3. Specifically stains mouse CCR3 transfectants but not the parental cell lines. Does not cross-react with Y3 cells transfected with human CCR3. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction