Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CCR3, Alexa Fluor 700, Clone: 83101, R&D Systems™
Specifications
Antigen | CCR3 |
---|---|
Clone | 83101 |
Dilution | Flow Cytometry 5 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB729N025
|
R&D Systems
FAB729N025 |
25 Tests | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
FAB729N100
|
R&D Systems
FAB729N100 |
100 Tests | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Host Species: Rat
Clone: 83101
Isotype: IgG2A
Species Reactivity: Ms
Immunogen: Y3 rat myeloid cell line transfected with mouse CCR3. Met1-Phe359
Applications:
Flow Cytometry
Specifications
CCR3 | |
Flow Cytometry 5 uL/10^6 cells | |
Monoclonal | |
Purified | |
Mouse | |
b-chemokine receptor, CC chemokine receptor 3, C-C chemokine receptor type 3, C-C CKR-3, CCR3, CCR-3, CD193, CD193 antigen, chemokine (C-C motif) receptor 3, CKR3CC-CKR-3CMKBR3CD193, eosinophil CC chemokine receptor 3, Eosinophil eotaxin receptor, MGC102841 | |
CCR3 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Detects mouse CCR3. Specifically stains mouse CCR3 transfectants but not the parental cell lines. Does not cross-react with Y3 cells transfected with human CCR3. |
83101 | |
Flow Cytometry | |
Alexa Fluor 700 | |
Rat | |
P51678 | |
1232 | |
Y3 rat myeloid cell line transfected with mouse CCR3, Met1-Phe359, Accession # P51678 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title