Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD104 Rat anti-Mouse, Unlabeled, Clone: 346-11A, BD

Catalog No. BDB553745 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553745 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553745 Supplier BD Biosciences Supplier No. 553745
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 346-11A antibody reacts with an epitope at the beginning of the cysteine-rich repeat region of the 200-kDa integrin β 4 chain (CD104), which is found on the cell surface as a heterodimeric complex with the integrin α 6 chain (CD49f). The α 6β 4 complex binds to laminins and is expressed on the basal surface of a variety of epithelial cell types, particularly on stratified squamous epithelia, and is also found in peripheral nerves, in certain subsets of endothelial cells, and on immature thymocytes. It has also been identified on a number of tumor tissues and participates in tumor progression events. Because of the localization of both human and mouse epidermal α 6β 4 integrin to hemidesmosomes, a role in epidermal adhesion to basement membrane has been suggested.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD104
Applications Flow Cytometry
Classification Monoclonal
Clone 346-11A
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Integrin β4 chain
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Tumor-associated antigen TSP-180 from a BALB/c mouse lung carcinoma
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.