Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11a (Integrin αL chain, LFA-1α) Mouse anti-Human, FITC, Clone: G-25.2, BD

Catalog No. BDB347983 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB347983 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB347983 Supplier BD Biosciences Supplier No. 347983
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD11a, clone G-25.2, is derived from hybridization of mouse Sp2/0 myeloma cells with spleen cells from BALB/c mice immunized with human cytotoxic T lymphocytes (CTL).

CD11a (Anti-LFA-1α) recognizes the Mr 180-kdalton (kDa) α chain of the leucocyte function–associated antigen-1 (LFA-1). LFA-1, a member of a subfamily of integrin receptors, is a leucocyte adhesion receptor that binds to an intercellular adhesion molecule (ICAM) on target tissues. This subfamily consists of three heterodimeric proteins that share a common noncovalently-associated β chain (CD18 antigen, Mr 95 kDa) but possess distinct α-subunits recognized by CD11a (anti-LFA-1α), CD11b (Leu-15), and CD11c (Leu-M5).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11a
Applications Flow Cytometry
Classification Monoclonal
Clone G-25.2
Concentration 6.25μg/mL
Conjugate FITC
Description LFA-1α
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Human cytotoxic T lymphocytes
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.